2-Chloro-N-cyclopropyl-4-fluoro-3-methoxybenzamide ≥97%
Supplier: AOBChem USA
|
Warning
|
46080-500MG
46080-1G
46080-5G
46080-25G
77327-046EA
120.51
USD
77327-046
77343-706
77363-354
77374-888
2-Chloro-N-cyclopropyl-4-fluoro-3-methoxybenzamide ≥97%
2-Chloro-N-cyclopropyl-4-fluoro-3-methoxybenzamide
Precaución: for Research Use Only
Formula:
C₁₁H₁₁ClFNO₂ MW: 243.662 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32902689 CAS Number: 2586126-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...