2-Chloro-N-cyclohexyl-4-fluoro-3-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

46079-500MG 46079-1G 46079-5G 46079-25G
77327-044EA 120.51 USD
77327-044 77343-704 77363-352 77374-886
2-Chloro-N-cyclohexyl-4-fluoro-3-methoxybenzamide ≥97%
2-Chloro-N-cyclohexyl-4-fluoro-3-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₇ClFNO₂
MW: 285.742 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902688
CAS Number: 2586126-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR