(2-Chloro-4-fluoro-3-methoxyphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

46078-500MG 46078-1G 46078-5G 46078-25G
77327-042EA 120.51 USD
77327-042 77343-702 77363-350 77374-884
(2-Chloro-4-fluoro-3-methoxyphenyl)(morpholino)methanone ≥97%
(2-Chloro-4-fluoro-3-methoxyphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClFNO₃
MW: 273.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902687
CAS Number: 2586127-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR