(2-Chloro-4-fluoro-3-methoxyphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

46077-500MG 46077-1G 46077-5G 46077-25G
77327-040EA 120.51 USD
77327-040 77343-700 77363-348 77374-882
(2-Chloro-4-fluoro-3-methoxyphenyl)(thiomorpholino)methanone ≥97%
(2-Chloro-4-fluoro-3-methoxyphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClFNO₂S
MW: 289.754 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902686
CAS Number: 2586127-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR