(2-Chloro-4-fluoro-3-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

46076-500MG 46076-1G 46076-5G 46076-25G
77327-038EA 120.51 USD
77327-038 77343-698 77363-346 77374-880
(2-Chloro-4-fluoro-3-methoxyphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(2-Chloro-4-fluoro-3-methoxyphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClFN₂O₂
MW: 286.73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902685
CAS Number: 2586126-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR