1,3-Dchloro-2-iodo-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

46069-500MG 46069-1G 46069-5G
77339-386EA 584.01 USD
77339-386 77350-632 77374-876
1,3-Dchloro-2-iodo-5-methoxybenzene ≥97%
1,3-Dchloro-2-iodo-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂IO
MW: 302.924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529958
CAS Number: 98273-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR