2-Bromo-1,3-dichloro-5-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

46066-1G 46066-5G 46066-25G
77340-676EA 60.77 USD
77340-676 77363-340 77374-874
2-Bromo-1,3-dichloro-5-ethoxybenzene ≥97%
2-Bromo-1,3-dichloro-5-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇BrCl₂O
MW: 269.951 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564091
CAS Number: 2168283-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR