3-(Benzyloxy)-4,6-dibromo-2-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

46062-500MG 46062-1G 46062-5G 46062-25G
77327-032EA 128.75 USD
77327-032 77344-240 77363-338 77374-872
3-(Benzyloxy)-4,6-dibromo-2-fluorobenzoic acid ≥95%
3-(Benzyloxy)-4,6-dibromo-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₉Br₂FO₃
MW: 404.026 g/mol
MDL Number: MFCD34756255
CAS Number: 2807471-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR