Ethyl 3,5-difluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

46051-500MG 46051-1G 46051-5G 46051-25G
77327-028EA 147.29 USD
77327-028 77345-264 77363-332 77385-990
Ethyl 3,5-difluoro-4-methoxybenzoate ≥97%
Ethyl 3,5-difluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111877
CAS Number: 1256482-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR