4-Bromo-2-((4-fluorobenzyl)oxy)-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

46045-1G 46045-5G 46045-25G
77341-314EA 82.4 USD
77341-314 77363-330 77374-868
4-Bromo-2-((4-fluorobenzyl)oxy)-1-methylbenzene ≥95%
4-Bromo-2-((4-fluorobenzyl)oxy)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902681
CAS Number: 1961405-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR