4-Chloro-5-fluoro-2-(trifluoromethyl)benzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
46013-500MG
46013-1G
46013-5G
77340-058EA
740.57
USD
77340-058
77350-630
77374-866
4-Chloro-5-fluoro-2-(trifluoromethyl)benzoic acid ≥97%
4-Chloro-5-fluoro-2-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₃ClF₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD28752590 CAS Number: 1807265-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...