[1,1'-Biphenyl]-2-carboxamide ≥97%

Supplier: AOBChem USA

Warning

45993-500MG 45993-1G 45993-5G 45993-25G
77327-024EA 140.08 USD
77327-024 77344-744 77363-324 77385-988
[1,1'-Biphenyl]-2-carboxamide ≥97%
[1,1'-Biphenyl]-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MW: 197.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075754
CAS Number: 13234-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR