N-Methyl-[1,1'-biphenyl]-2-carboxamide ≥97%

Supplier: AOBChem USA

Warning

45992-500MG 45992-1G 45992-5G 45992-25G
77327-022EA 121.54 USD
77327-022 77343-864 77363-322 77374-860
N-Methyl-[1,1'-biphenyl]-2-carboxamide ≥97%
N-Methyl-[1,1'-biphenyl]-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO
MW: 211.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04216169
CAS Number: 29585-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR