1-(3-Chloro-2-fluoropyridin-4-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

45973-500MG 45973-1G 45973-5G
77338-516EA 494.4 USD
77338-516 77350-628 77374-858
1-(3-Chloro-2-fluoropyridin-4-yl)ethanone ≥97%
1-(3-Chloro-2-fluoropyridin-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅ClFNO
MW: 173.572 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257920
CAS Number: 1256834-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR