1-(3-Chloro-2-fluoropyridin-4-yl)ethanol ≥95%

Supplier: AOBChem USA

Warning

45972-500MG 45972-1G 45972-5G 45972-25G
77327-020EA 257.5 USD
77327-020 77350-626 77363-320 77385-986
1-(3-Chloro-2-fluoropyridin-4-yl)ethanol ≥95%
1-(3-Chloro-2-fluoropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₇ClFNO
Storage Temperature: Ambient
MDL Number: MFCD28138765
CAS Number: 1149586-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR