1-(5-Bromo-2-chloropyridin-4-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

45965-500MG 45965-1G 45965-5G
77337-648EA 434.66 USD
77337-648 77350-624 77374-856
1-(5-Bromo-2-chloropyridin-4-yl)ethanol ≥97%
1-(5-Bromo-2-chloropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₇BrClNO
Storage Temperature: Ambient
MDL Number: MFCD28346787
CAS Number: 1823421-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR