Methyl 2,4-dibromo-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

45948-500MG 45948-1G 45948-5G
77337-854EA 449.08 USD
77337-854 77350-622 77374-854
Methyl 2,4-dibromo-5-methoxybenzoate ≥97%
Methyl 2,4-dibromo-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈Br₂O₃
MW: 323.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707151
CAS Number: 875246-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR