(3-Bromo-5-fluorophenyl)(thiomorpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

45939-500MG 45939-1G 45939-5G 45939-25G
77327-014EA 99.91 USD
77327-014 77342-790 77363-312 77374-848
(3-Bromo-5-fluorophenyl)(thiomorpholino)methanone ≥95%
(3-Bromo-5-fluorophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrFNOS
MW: 304.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564090
CAS Number: 2734774-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR