3-Bromo-N-cyclohexyl-5-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

45937-500MG 45937-1G 45937-5G 45937-25G
77327-012EA 87.55 USD
77327-012 77342-298 77363-310 77374-846
3-Bromo-N-cyclohexyl-5-fluorobenzamide ≥97%
3-Bromo-N-cyclohexyl-5-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrFNO
MW: 300.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20146155
CAS Number: 1329464-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR