(6-Bromo-3-ethoxy-2-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

45935-500MG 45935-1G 45935-5G 45935-25G
77327-010EA 140.08 USD
77327-010 77344-710 77363-308 77385-982
(6-Bromo-3-ethoxy-2-fluorophenyl)methanol ≥97%
(6-Bromo-3-ethoxy-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD32707148
CAS Number: 2386997-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR