Methyl 3-bromo-2-ethoxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

45924-500MG 45924-1G 45924-25G 45924-5G
77327-006EA 285.31 USD
77327-006 77350-616 77385-978 77374-842
Methyl 3-bromo-2-ethoxy-5-methylbenzoate ≥97%
Methyl 3-bromo-2-ethoxy-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₃
MW: 273.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564089
CAS Number: 1527969-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR