3-Bromo-2,4,6-trimethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

45914-500MG 45914-1G 45914-5G 45914-25G
77327-002EA 223.51 USD
77327-002 77350-612 77363-304 77385-974
3-Bromo-2,4,6-trimethylbenzoic acid ≥95%
3-Bromo-2,4,6-trimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107207
CAS Number: 5333-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR