(3-Ethoxy-2-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

45911-500MG 45911-1G 45911-5G 45911-25G
77326-998EA 202.91 USD
77326-998 77350-608 77363-300 77385-970
(3-Ethoxy-2-fluorophenyl)methanol ≥97%
(3-Ethoxy-2-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707139
CAS Number: 2432848-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR