Methyl 3-ethoxy-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

45910-500MG 45910-1G 45910-5G 45910-25G
77326-996EA 208.06 USD
77326-996 77350-606 77363-298 77385-968
Methyl 3-ethoxy-2-fluorobenzoate ≥97%
Methyl 3-ethoxy-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD27578379
CAS Number: 1820604-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR