4-Chloro-2-fluoro-3-formyl-5-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
45905-500MG
45905-1G
45905-5G
45905-25G
77326-994EA
202.91
USD
77326-994
77350-604
77363-296
77385-966
4-Chloro-2-fluoro-3-formyl-5-methylbenzoic acid ≥97%
4-Chloro-2-fluoro-3-formyl-5-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₆ClFO₃ MW: 216.594 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD33403812 CAS Number: 2624417-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...