2-Chloro-1-fluoro-3-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

45777-500MG 45777-1G 45777-5G
77339-432EA 590.19 USD
77339-432 77350-598 77374-836
2-Chloro-1-fluoro-3-methoxy-5-methylbenzene ≥97%
2-Chloro-1-fluoro-3-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClFO
MW: 174.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751725
CAS Number: 1805041-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR