3-Bromo-2-chloro-5-methylanisole ≥95%

Supplier: AOBChem USA

Warning

45776-500MG 45776-1G 45776-5G
77335-534EA 357.41 USD
77335-534 77350-596 77374-834
3-Bromo-2-chloro-5-methylanisole ≥95%
3-Bromo-2-chloro-5-methylanisole

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD12547794
CAS Number: 1208075-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR