6-(3,4-Dichlorophenyl)nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

45699-500MG 45699-1G 45699-5G
77338-242EA 470.71 USD
77338-242 77350-594 77374-832
6-(3,4-Dichlorophenyl)nicotinaldehyde ≥97%
6-(3,4-Dichlorophenyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₇Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD04115473
CAS Number: 898796-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR