6-(4-hydroxyphenyl)picolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

45678-500MG 45678-1G 45678-5G
77336-364EA 387.28 USD
77336-364 77350-590 77374-828
6-(4-hydroxyphenyl)picolinaldehyde ≥97%
6-(4-hydroxyphenyl)picolinaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD24687682
CAS Number: 1367126-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR