(E)-ethyl 3-(2-chloro-4-fluorophenyl)acrylate ≥97%

Supplier: AOBChem USA

Warning

45636-500MG 45636-1G 45636-5G 45636-25G
77326-978EA 245.14 USD
77326-978 77350-582 77363-284 77385-956
(E)-ethyl 3-(2-chloro-4-fluorophenyl)acrylate ≥97%
(E)-ethyl 3-(2-chloro-4-fluorophenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀ClFO₂
MW: 228.647 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22124751
CAS Number: 1257121-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR