8-Bromo-5,6-difluoroquinoline ≥97%

Supplier: AOBChem USA

Warning

45629-500MG 45629-1G 45629-25G 45629-5G
77326-976EA 299.73 USD
77326-976 77350-580 77385-954 77374-820
8-Bromo-5,6-difluoroquinoline ≥97%
8-Bromo-5,6-difluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₄BrF₂N
MW: 244.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564083
CAS Number: 225366-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR