2-(2-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

45622-500MG 45622-1G 45622-25G 45622-5G
77326-974EA 298.7 USD
77326-974 77350-578 77385-952 77374-818
2-(2-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-((4-Bromo-2-fluorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₉H₂₁BBrFO₃
Storage Temperature: Ambient
CAS Number: 2246573-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR