2-(3-Bromo-4-fluorophenyl)-2-oxoethyl acetate ≥97%

Supplier: AOBChem USA

Warning

45578-500MG 45578-1G 45578-5G 45578-25G
77326-970EA 120.51 USD
77326-970 77343-694 77363-280 77374-814
2-(3-Bromo-4-fluorophenyl)-2-oxoethyl acetate ≥97%
2-(3-Bromo-4-fluorophenyl)-2-oxoethyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD32642074
CAS Number: 2734779-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR