2-Oxopropyl 4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

45575-500MG 45575-1G 45575-5G 45575-25G
77326-968EA 90.64 USD
77326-968 77342-386 77363-278 77374-812
2-Oxopropyl 4-methoxybenzoate ≥97%
2-Oxopropyl 4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05106639
CAS Number: 101395-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR