2-Oxopropyl 4-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

45573-1G 45573-5G 45573-25G
77341-566EA 93.73 USD
77341-566 77363-274 77374-808
2-Oxopropyl 4-chlorobenzoate ≥97%
2-Oxopropyl 4-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD05106247
CAS Number: 900937-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR