1-Fluoro-3-isopropoxy-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

45569-1G 45569-5G 45569-25G
77341-310EA 82.4 USD
77341-310 77363-270 77374-804
1-Fluoro-3-isopropoxy-5-methoxybenzene ≥97%
1-Fluoro-3-isopropoxy-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD23114330
CAS Number: 1369873-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR