1-(Difluoromethoxy)-3-fluoro-5-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

45568-500MG 45568-1G 45568-5G 45568-25G
77326-966EA 182.31 USD
77326-966 77346-950 77363-268 77385-950
1-(Difluoromethoxy)-3-fluoro-5-methoxybenzene ≥97%
1-(Difluoromethoxy)-3-fluoro-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇F₃O₂
MW: 192.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394920
CAS Number: 1261760-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR