1-(Benzyloxy)-3-fluoro-5-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

45567-1G 45567-5G 45567-25G
77341-308EA 82.4 USD
77341-308 77363-266 77374-802
1-(Benzyloxy)-3-fluoro-5-methoxybenzene ≥95%
1-(Benzyloxy)-3-fluoro-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707131
CAS Number: 2432849-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR