6-Bromo-2-methoxynicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

45554-500MG 45554-1G 45554-5G 45554-25G
77326-964EA 189.52 USD
77326-964 77350-576 77363-264 77385-948
6-Bromo-2-methoxynicotinaldehyde ≥95%
6-Bromo-2-methoxynicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188724
CAS Number: 58819-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR