(5-Bromo-2,4-difluoro-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

45552-500MG 45552-1G 45552-5G 45552-25G
77326-962EA 98.88 USD
77326-962 77342-700 77363-262 77374-800
(5-Bromo-2,4-difluoro-3-methylphenyl)methanol ≥97%
(5-Bromo-2,4-difluoro-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707128
CAS Number: 2432848-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR