Methyl 5-bromo-2,4-difluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

45551-500MG 45551-1G 45551-5G 45551-25G
77326-960EA 87.55 USD
77326-960 77342-296 77363-260 77374-798
Methyl 5-bromo-2,4-difluoro-3-methylbenzoate ≥97%
Methyl 5-bromo-2,4-difluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32707127
CAS Number: 2432849-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR