5-Bromo-2,4-difluoro-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

45549-500MG 45549-1G 45549-5G 45549-25G
77326-958EA 120.51 USD
77326-958 77343-692 77363-256 77374-794
5-Bromo-2,4-difluoro-3-methylphenol ≥97%
5-Bromo-2,4-difluoro-3-methylphenol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD32902678
CAS Number: 2586126-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR