Ethyl 6-fluoro-2-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

45545-500MG 45545-1G 45545-5G
77326-956EA 342.99 USD
77326-956 77350-574 77374-792
Ethyl 6-fluoro-2-methoxy-3-methylbenzoate ≥97%
Ethyl 6-fluoro-2-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD32707124
CAS Number: 2112934-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR