(2,4-Dichloro-6-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

45531-1G 45531-5G 45531-25G
77341-962EA 109.18 USD
77341-962 77363-248 77374-786
(2,4-Dichloro-6-fluorophenyl)(methyl)sulfane ≥97%
(2,4-Dichloro-6-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FS
MW: 211.084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786339
CAS Number: 1806277-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR