Methyl 3-bromo-2,4-dichloro-6-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

45530-500MG 45530-1G 45530-5G 45530-25G
77326-946EA 151.41 USD
77326-946 77345-422 77363-246 77385-940
Methyl 3-bromo-2,4-dichloro-6-fluorobenzoate ≥97%
Methyl 3-bromo-2,4-dichloro-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₄BrCl₂FO₂
MW: 301.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403808
CAS Number: 2624417-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR