(2,6-Dichloro-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

45500-1G 45500-5G 45500-25G
77341-270EA 80.34 USD
77341-270 77363-242 77374-782
(2,6-Dichloro-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2,6-Dichloro-3-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₅Cl₂F₃S
Storage Temperature: Ambient
MDL Number: MFCD32707122
CAS Number: 2432848-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR