2-Bromo-1,3-dichloro-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

45499-500MG 45499-1G 45499-5G
77335-800EA 366.68 USD
77335-800 77350-568 77374-780
2-Bromo-1,3-dichloro-4-(trifluoromethyl)benzene ≥97%
2-Bromo-1,3-dichloro-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂BrCl₂F₃
Storage Temperature: Ambient
MDL Number: MFCD29919823
CAS Number: 859794-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR