Methyl 2,6-dichloro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

45490-500MG 45490-1G 45490-5G 45490-25G
77326-944EA 130.81 USD
77326-944 77344-334 77363-240 77385-938
Methyl 2,6-dichloro-3-(trifluoromethyl)benzoate ≥97%
Methyl 2,6-dichloro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₅Cl₂F₃O₂
MW: 273.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403806
CAS Number: 2385055-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR