4-Bromo-2-chloro-5-(trifluoromethyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

45473-500MG 45473-1G 45473-5G 45473-25G
77326-938EA 224.54 USD
77326-938 77350-562 77363-230 77385-934
4-Bromo-2-chloro-5-(trifluoromethyl)benzonitrile ≥97%
4-Bromo-2-chloro-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₂BrClF₃N
Storage Temperature: Ambient
MDL Number: MFCD34756218
CAS Number: 2167638-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR