2-Fluoro-3-methyl-6-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

45437-500MG 45437-1G 45437-5G 45437-25G
77326-926EA 97.85 USD
77326-926 77342-658 77363-218 77374-760
2-Fluoro-3-methyl-6-propoxybenzoic acid ≥97%
2-Fluoro-3-methyl-6-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403797
CAS Number: 2624417-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR